4-Amino-5-methyl-3-nitropyridine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-5-nitropyridin-4-amine

OR345284-250MG
APOSOR345284-250MGEA 63.8 EUR
APOSOR345284-250MG
4-Amino-5-methyl-3-nitropyridine 97%
4-Amino-5-methyl-3-nitropyridine
Formel: C₆H₇N₃O₂
Molecular Weight: 153,139 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11656245
CAS-Nummer: 18227-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR