2-Iodopyridin-3-amine 96%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-iodopyridine

OR345293-1G
APOSOR345293-1GEA 108 EUR
APOSOR345293-1G
2-Iodopyridin-3-amine 96%
2-Iodopyridin-3-amine
Formel: C₅H₅IN₂
Molecular Weight: 220.0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779506
CAS-Nummer: 209286-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR