4-Iodo-2-methylpyridine 98%

Lieferant: Apollo Scientific

OR345298-1G
APOSOR345298-1GEA 133 EUR
APOSOR345298-1G
4-Iodo-2-methylpyridine 98%
4-Iodo-2-methylpyridine
Formel: C₆H₆IN
Molecular Weight: 219,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702484
CAS-Nummer: 22282-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR