2,4-Dibromo-3-hydroxy-6-methylpyridine 95+%

Lieferant: Apollo Scientific

Synonyme: 2,4-Dibromo-6-methylpyridin-3-ol

OR345302-1G
APOSOR345302-1GEA 66.7 EUR
APOSOR345302-1G
2,4-Dibromo-3-hydroxy-6-methylpyridine 95+%
2,4-Dibromo-3-hydroxy-6-methylpyridine
Formel: C₆H₅Br₂NO
Molecular Weight: 266,918 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09753265
CAS-Nummer: 23003-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR