3-Methoxy-2-methyl-6-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR345305-1G
APOSOR345305-1GEA 520 EUR
APOSOR345305-1G
3-Methoxy-2-methyl-6-nitropyridine
3-Methoxy-2-methyl-6-nitropyridine
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839293
CAS-Nummer: 23904-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR