5-Amino-2-cyano-3-methylpyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-3-methylpicolinonitrile
OR345307-1G
OR345307-5G
APOSOR345307-1GEA
143
EUR
APOSOR345307-1G
APOSOR345307-5G
5-Amino-2-cyano-3-methylpyridine
5-Amino-2-cyano-3-methylpyridine
Formel:
C₇H₇N₃ Molecular Weight: 133.151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375122 CAS-Nummer: 252056-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...