5-Amino-2-cyano-3-methylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-methylpicolinonitrile

OR345307-1G OR345307-5G
APOSOR345307-1GEA 143 EUR
APOSOR345307-1G APOSOR345307-5G
5-Amino-2-cyano-3-methylpyridine
5-Amino-2-cyano-3-methylpyridine
Formel: C₇H₇N₃
Molecular Weight: 133.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375122
CAS-Nummer: 252056-70-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR