4-Amino-2-methoxy-3-nitropyridine 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-3-nitropyridin-4-amine

OR345322-250MG
APOSOR345322-250MGEA 79.9 EUR
APOSOR345322-250MG
4-Amino-2-methoxy-3-nitropyridine 98%
4-Amino-2-methoxy-3-nitropyridine
Formel: C₆H₇N₃O₃
Molecular Weight: 169,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437848
CAS-Nummer: 33623-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR