2-Amino-5-chloropyridin-3-ol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-chloro-3-hydroxypyridine, 2-Amino-3-hydroxy-5-chloropyridine

OR345338-25G
APOSOR345338-25GEA 94.4 EUR
APOSOR345338-25G
2-Amino-5-chloropyridin-3-ol 98%
2-Amino-5-chloropyridin-3-ol
Formel: C₅H₅ClN₂O
Molecular Weight: 144.5590 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706179
CAS-Nummer: 40966-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR