2-Brom-p-toluonitril 98%

Lieferant: Apollo Scientific

OR345343-100G
APOSOR345343-100GEA 121 EUR
APOSOR345343-100G
2-Brom-p-toluonitril 98%
2-Brom-p-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00158968
CAS-Nummer: 42872-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR