2-Amino-6-chlor-4-methylpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-4-methylpyridin-2-amine

OR345359-1G OR345359-5G OR345359-10G
APOSOR345359-1GEA 89.9 EUR
APOSOR345359-1G APOSOR345359-5G APOSOR345359-10G
2-Amino-6-chlor-4-methylpyridin 97%
2-Amino-6-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.5862 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839256
CAS-Nummer: 51564-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR