2-Amino-6-chlor-4-methylpyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-4-methylpyridin-2-amine
OR345359-1G
OR345359-5G
OR345359-10G
APOSOR345359-1GEA
89.9
EUR
APOSOR345359-1G
APOSOR345359-5G
APOSOR345359-10G
2-Amino-6-chlor-4-methylpyridin 97%
2-Amino-6-chloro-4-methylpyridine
Formel:
C₆H₇ClN₂ Molecular Weight: 142.5862 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839256 CAS-Nummer: 51564-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...