3-Chloro-5-hydroxybenzoic acid
Lieferant: Apollo Scientific
OR345363-1G
APOSOR345363-1GEA
48
EUR
APOSOR345363-1G
3-Chloro-5-hydroxybenzoic acid
3-Chloro-5-hydroxybenzoic acid
Formel:
C₇H₅ClO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114327 CAS-Nummer: 53984-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...