2-Bromo-5-hydroxy-3-methylpyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-hydroxy-3-picoline

OR345364-5G
APOSOR345364-5GEA 116 EUR
APOSOR345364-5G
2-Bromo-5-hydroxy-3-methylpyridine 98%
2-Bromo-5-hydroxy-3-methylpyridine
Formel: C₆H₆BrNO
Molecular Weight: 188.024 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839267
CAS-Nummer: 1003711-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR