2-Amino-3-chloro-4-methylpyridine ≥97%

Lieferant: Apollo Scientific

OR345371-250MG
APOSOR345371-250MGEA 140 EUR
APOSOR345371-250MG
2-Amino-3-chloro-4-methylpyridine ≥97%
2-Amino-3-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142,586 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09839234
CAS-Nummer: 56960-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR