5-Bromo-3-chloropyridin-2(1H)-one 98%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-3-chloro-2-hydroxypyridine
OR345375-25G
APOSOR345375-25GEA
85.7
EUR
APOSOR345375-25G
5-Bromo-3-chloropyridin-2(1H)-one 98%
5-Bromo-3-chloropyridin-2(1H)-one
Formel:
C₅H₃BrClNO Molecular Weight: 208.44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662389 CAS-Nummer: 58236-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...