5-Chloro-2-hydroxy-3-methylpyridine
Lieferant: Apollo Scientific
Synonyme:
5-Chloro-3-methylpyridin-2-ol
OR345376-1G
APOSOR345376-1GEA
74.1
EUR
APOSOR345376-1G
5-Chloro-2-hydroxy-3-methylpyridine
5-Chloro-2-hydroxy-3-methylpyridine
Formel:
C₆H₆ClNO Molecular Weight: 143,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126916 CAS-Nummer: 58498-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...