2-Cyan-4-methylanilin 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-methylbenzonitril, 2-Amino-5-methylbenzoesäurenitril

OR345378-5G
APOSOR345378-5GEA 104 EUR
APOSOR345378-5G
2-Cyan-4-methylanilin 98%
2-Cyan-4-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00267506
CAS-Nummer: 5925-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR