3-Methoxy-5-methylbenzoic acid 97%

Lieferant: Apollo Scientific

OR345381-5G
APOSOR345381-5GEA 145 EUR
APOSOR345381-5G
3-Methoxy-5-methylbenzoic acid 97%
3-Methoxy-5-methylbenzoic acid
Formel: C9H10O3
Molecular Weight: 166,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405314
CAS-Nummer: 62089-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR