3-Bromo-2-hydroxy-5-iodopyridine 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-iodopyridin-2-ol

OR345385-1G
APOSOR345385-1GEA 52.2 EUR
APOSOR345385-1G
3-Bromo-2-hydroxy-5-iodopyridine 95+%
3-Bromo-2-hydroxy-5-iodopyridine
Formel: C₅H₃BrINO
Molecular Weight: 299,892 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044276
CAS-Nummer: 637348-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR