5-Brom-4-chlor-2-(methylsulfanyl)pyrimidin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine
OR345386-25G
APOSOR345386-25GEA
101
EUR
APOSOR345386-25G
5-Brom-4-chlor-2-(methylsulfanyl)pyrimidin
5-Bromo-4-chloro-2-(methylthio)pyrimidine
Formel:
C₅H₄BrClN₂S Molecular Weight: 239,5200 g/mol Schmelzpunkt: 45…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023232 CAS-Nummer: 63810-78-6 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...