5-Methyl-2-nitrobenzonitrile 95+%

Lieferant: Apollo Scientific

OR345387-5G
APOSOR345387-5GEA 72.5 EUR
APOSOR345387-5G
5-Methyl-2-nitrobenzonitrile 95+%
5-Methyl-2-nitrobenzonitrile
Formel: C8H6N2O2
Molecular Weight: 162,1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00267507
CAS-Nummer: 64113-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR