2-Amino-5-bromo-6-methyl-3-nitropyridine 97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-6-methyl-3-nitropyridin-2-amine

OR345393-5G
APOSOR345393-5GEA 120 EUR
APOSOR345393-5G
2-Amino-5-bromo-6-methyl-3-nitropyridine 97%
2-Amino-5-bromo-6-methyl-3-nitropyridine
Formel: C₆H₆BrN₃O₂
Molecular Weight: 232,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091828
CAS-Nummer: 68957-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR