2-Amino-5-ethoxy-6-methylpyridine 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Ethoxy-6-methylpyridin-2-amine

OR345403-250MG
APOSOR345403-250MGEA 104 EUR
APOSOR345403-250MG
2-Amino-5-ethoxy-6-methylpyridine 95+%
2-Amino-5-ethoxy-6-methylpyridine
Formel: C₈H₁₂N₂O
Molecular Weight: 152.194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000613
CAS-Nummer: 73101-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR