3-Bromo-2-isopropoxy-5-methylpyridine 97%

Lieferant: Apollo Scientific

OR345408-1G
APOSOR345408-1GEA 88.6 EUR
APOSOR345408-1G
3-Bromo-2-isopropoxy-5-methylpyridine 97%
3-Bromo-2-isopropoxy-5-methylpyridine
Formel: C₉H₁₂BrNO
Molecular Weight: 230.102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17926523
CAS-Nummer: 760207-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR