2-Chlor-3-methoxy-6-nitropyridin 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-3-methoxy-6-nitropyridine

OR345409-1G
APOSOR345409-1GEA 114 EUR
APOSOR345409-1G
2-Chlor-3-methoxy-6-nitropyridin 95+%
2-Chlor-3-methoxy-6-nitropyridin
Formel: C₆H₅BrN₂O₃
Molecular Weight: 233,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06662266
CAS-Nummer: 76066-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR