2-Bromo-6-methoxyisonicotinic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-6-methoxypyridine-4-carboxylic acid

OR345428-250MG
APOSOR345428-250MGEA 121 EUR
APOSOR345428-250MG
2-Bromo-6-methoxyisonicotinic acid 97%
2-Bromo-6-methoxyisonicotinic acid
Formel: C₇H₆BrNO₃
Molecular Weight: 232,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101033
CAS-Nummer: 853029-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR