2-Bromo-3-chloro-4-methylpyridine 98%
Lieferant: Apollo Scientific
OR345441-5G
APOSOR345441-5GEA
71.2
EUR
APOSOR345441-5G
2-Bromo-3-chloro-4-methylpyridine 98%
2-Bromo-3-chloro-4-methylpyridine
Formel:
C₆H₅BrClN Molecular Weight: 206.468 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277283 CAS-Nummer: 884495-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...