6-Hydroxy-3-iodo-2-methylpyridine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Iodo-6-methylpyridin-2(1H)-one

OR345454-1G
APOSOR345454-1GEA 58 EUR
APOSOR345454-1G
6-Hydroxy-3-iodo-2-methylpyridine 98%
6-Hydroxy-3-iodo-2-methylpyridine
Formel: C₆H₆INO
Molecular Weight: 235,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037468
CAS-Nummer: 927870-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR