5-Nitropyridin-3-amine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-nitropyridine

OR345456-1G
APOSOR345456-1GEA 114 EUR
APOSOR345456-1G
5-Nitropyridin-3-amine 98%
5-Nitropyridin-3-amine
Formel: C5H5N3O2
Molecular Weight: 139.1121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114212
CAS-Nummer: 934-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR