2-Amino-5-bromo-4-chloro-3-nitropyridine 97%
Lieferant: Apollo Scientific
OR345460-5G
APOSOR345460-5GEA
149
EUR
APOSOR345460-5G
2-Amino-5-bromo-4-chloro-3-nitropyridine 97%
2-Amino-5-bromo-4-chloro-3-nitropyridine
Formel:
C₅H₃BrClN₃O₂ Molecular Weight: 252,4500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110693 CAS-Nummer: 942947-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...