5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-3-chloro-7-azaindole
OR345464-5G
APOSOR345464-5GEA
101
EUR
APOSOR345464-5G
5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine
5-Bromo-3-chloro-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₄BrClN₂ Molecular Weight: 231,4771 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817420 CAS-Nummer: 951626-91-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...