2,6-Dichloro-3-methyl-5-nitropyridine 95+%

Lieferant: Apollo Scientific

OR345475-1G
APOSOR345475-1GEA 77 EUR
APOSOR345475-1G
2,6-Dichloro-3-methyl-5-nitropyridine 95+%
2,6-Dichloro-3-methyl-5-nitropyridine
Formel: C₆H₄Cl₂N₂O₂
Molecular Weight: 207,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014968
CAS-Nummer: 58596-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR