2-Amino-6-bromo-3-methoxypyridine 97%

Lieferant: Apollo Scientific

OR345483-1G OR345483-250MG OR345483-5G
APOSOR345483-1GEA 159 EUR
APOSOR345483-1G APOSOR345483-250MG APOSOR345483-5G
2-Amino-6-bromo-3-methoxypyridine 97%
2-Amino-6-bromo-3-methoxypyridine
Formel: C₆H₇BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD15142726
CAS-Nummer: 916737-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR