5-Bromo-2-ethoxypyridin-3-amine 97%
Lieferant: Apollo Scientific
OR345490-250MG
APOSOR345490-250MGEA
66.7
EUR
APOSOR345490-250MG
5-Bromo-2-ethoxypyridin-3-amine 97%
5-Bromo-2-ethoxypyridin-3-amine
Formel:
C₇H₉BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375021 CAS-Nummer: 886373-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...