2-Bromo-6-methoxy-3-nitropyridine

Lieferant: Apollo Scientific

OR345495-5G
APOSOR345495-5GEA 58 EUR
APOSOR345495-5G
2-Bromo-6-methoxy-3-nitropyridine
2-Bromo-6-methoxy-3-nitropyridine
Formel: C₆H₅BrN₂O₃
Molecular Weight: 233.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774106
CAS-Nummer: 344296-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR