2-Chloro-5-hydroxy-3-methylpyridine 96%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-5-methyl-3-pyridinol, 6-Chloro-5-methylpyridin-3-ol

OR345498-5G
APOSOR345498-5GEA 212 EUR
APOSOR345498-5G
2-Chloro-5-hydroxy-3-methylpyridine 96%
2-Chloro-5-hydroxy-3-methylpyridine
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519295
CAS-Nummer: 54232-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR