2-Chloro-5-methoxy-3-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR345499-100MG
APOSOR345499-100MGEA 148 EUR
APOSOR345499-100MG
2-Chloro-5-methoxy-3-nitropyridine
2-Chloro-5-methoxy-3-nitropyridine
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835251
CAS-Nummer: 1003711-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR