3-Amino-5-bromo-2-iodopyridine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-iodopyridin-3-amine

OR345518-250MG
APOSOR345518-250MGEA 109 EUR
APOSOR345518-250MG
3-Amino-5-bromo-2-iodopyridine 98%
3-Amino-5-bromo-2-iodopyridine
Formel: C₅H₄BrIN₂
Molecular Weight: 298,907 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014978
CAS-Nummer: 1180678-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR