3-Bromo-5-(prop-1-en-2-yl)pyridine 95+%
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-5-isopropenylpyridine
OR345529-250MG
APOSOR345529-250MGEA
174
EUR
APOSOR345529-250MG
3-Bromo-5-(prop-1-en-2-yl)pyridine 95+%
3-Bromo-5-(prop-1-en-2-yl)pyridine
Formel:
C₈H₈BrN Molecular Weight: 198,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19689077 CAS-Nummer: 118775-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...