4-Amino-3-bromo-5-chloropyridine 98%
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-5-chloropyridin-4-amine
OR345543-5G
APOSOR345543-5GEA
112
EUR
APOSOR345543-5G
4-Amino-3-bromo-5-chloropyridine 98%
4-Amino-3-bromo-5-chloropyridine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,456 g/mol Schmelzpunkt: 160…170 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10686645 CAS-Nummer: 159783-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...