4-Chloro-2-methoxy-3-nitropyridine 97%

Lieferant: Apollo Scientific

OR345550-250MG
APOSOR345550-250MGEA 235 EUR
APOSOR345550-250MG
4-Chloro-2-methoxy-3-nitropyridine 97%
4-Chloro-2-methoxy-3-nitropyridine
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014990
CAS-Nummer: 934180-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR