1-Methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid 97%

Lieferant: Apollo Scientific

OR345604-25G
APOSOR345604-25GEA 149 EUR
APOSOR345604-25G
1-Methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid 97%
1-Methyl-2-oxo-1,2-dihydropyridine-4-carboxylic acid
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701415
CAS-Nummer: 33972-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR