2-Methyl-2-propanyl-(6-aminohexyl)carbamat 97%

Lieferant: Apollo Scientific

Synonyme: N-tert-Butoxycarbonyl-1,6-hexanediamine, N-Boc-1,6-Diaminohexane

OR345618-25G OR345618-100G
APOSOR345618-25GEA 81.2 EUR
APOSOR345618-25G APOSOR345618-100G
2-Methyl-2-propanyl-(6-aminohexyl)carbamat 97%
N-Boc-1,6-diaminohexane
Formel: C₁₁H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00671489
CAS-Nummer: 51857-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR