4-[(Cyclopropylcarbamoyl)amino]benzeneboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Cyclopropyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea, 4-(3-Cyclopropylureido)benzeneboronic acid, pinacol ester

OR3457-1G
APOSOR3457-1GEA 232 EUR
APOSOR3457-1G
4-[(Cyclopropylcarbamoyl)amino]benzeneboronic acid, pinacol ester 97%
4-[(Cyclopropylcarbamoyl)amino]benzeneboronic acid, pinacol ester
Formel: C₁₆H₂₃BN₂O₃
Schmelzpunkt: 227…229 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689493
CAS-Nummer: 874297-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR