Methyl 2-bromooxazole-5-carboxylate

Lieferant: Apollo Scientific

OR345709-1G
APOSOR345709-1GEA 763 EUR
APOSOR345709-1G
Methyl 2-bromooxazole-5-carboxylate
Methyl 2-bromooxazole-5-carboxylate
Formel: C₅H₄BrNO₃
Molecular Weight: 205.994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109415
CAS-Nummer: 1092351-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR