5-Iodo-2-methoxypyrimidine 95%
Lieferant: Apollo Scientific
OR345710-1G
APOSOR345710-1GEA
236
EUR
APOSOR345710-1G
5-Iodo-2-methoxypyrimidine 95%
5-Iodo-2-methoxypyrimidine
Formel:
C₅H₅IN₂O Molecular Weight: 236,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18412677 CAS-Nummer: 101803-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...