(2-Aminopyrimidin-5-yl)methanol

Lieferant: Apollo Scientific

OR345712-1G
APOSOR345712-1GEA 172 EUR
APOSOR345712-1G
(2-Aminopyrimidin-5-yl)methanol
(2-Aminopyrimidin-5-yl)methanol
Formel: C₅H₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09991890
CAS-Nummer: 120747-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR