4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile

Lieferant: Apollo Scientific

OR345713-1G
APOSOR345713-1GEA 200 EUR
APOSOR345713-1G
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile
Formel: C₇H₆ClN₃S
Molecular Weight: 199,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28134480
CAS-Nummer: 1208170-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR