2-(Chloromethyl)-4,6-dimethylpyrimidine

Lieferant: Apollo Scientific

OR345718-1G
APOSOR345718-1GEA 351 EUR
APOSOR345718-1G
2-(Chloromethyl)-4,6-dimethylpyrimidine
2-(Chloromethyl)-4,6-dimethylpyrimidine
Formel: C₇H₉ClN₂
Molecular Weight: 156,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864267
CAS-Nummer: 74502-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR